EMAIL THIS PAGE TO A FRIEND
Mutagenesis
Toxicogenomics analysis of mouse lung responses following exposure to titanium dioxide nanomaterials reveal their disease potential at high doses.
PMID 27760801
Abstract
Titanium dioxide nanoparticles (TiO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N2040
2-NP, ≥98% (HPLC)
C14H10N2O