EMAIL THIS PAGE TO A FRIEND
Amino acids
β-Conglycinin enhances autophagy in porcine enterocytes.
PMID 27761755
Abstract
β-Conglycinin (β-CG) is well known for inducing intestinal allergies and dysfunction in neonates and young pigs. However, the underlying mechanisms are largely unknown. In this study, to clarify the role of autophagy in β-CG-induced cytotoxicity, we investigated the effects of β-CG on cell viability and autophagy activity in porcine enterocytes (IPEC-1 cells). The results indicated that the cell viability was decreased with the increasing levels of β-CG. β-CG treatment enhanced the eGFP-LC3 puncta per cells and LC3-II/LC3-I, and the latter was further increased in IPEC-1 cells cultured with bafilomycin A1. We conclude that β-CG enhances autophagy activity in enterocytes.