EMAIL THIS PAGE TO A FRIEND
Neurotoxicity research
The Presence of the Neuronal Nitric Oxide Synthase Isoform in the Intervertebral Disk.
PMID 27761804
Abstract
Intervertebral disk degeneration is a progressive and debilitating disease with multifactorial causes. Nitric oxide (NO) might contribute to the cell death pathway. We evaluated the presence of the constitutive form of the neuronal NOS (nNOS) in both health and degenerated intervertebral disk through qPCR and immunohistochemistry. We also analyzed the potential role of nNOS modulation in the tail needle puncture model of intervertebral disk degeneration. Male Wistar rats were submitted to percutaneous disk puncture with a 21-gauge needle of coccygeal vertebras. The selective nNOS pharmacological inhibitor N