EMAIL THIS PAGE TO A FRIEND
Stem cells and development
Sutures Possess Strong Regenerative Capacity for Calvarial Bone Injury.
PMID 27762665
Abstract
Repair of calvarial bony defects remains challenging for craniofacial surgeons. Injury experiments on animal calvarial bones are widely used to study healing mechanisms and test tissue engineering approaches. Previously, we identified Gli1+ cells within the calvarial sutures as stem cells supporting calvarial bone turnover and injury repair. In this study, we tested the regenerative capacity of the suture region compared with other areas of calvarial bone. Injuries were made to mouse sagittal sutures or other areas of the calvarial bone at varying distances from the suture. Samples were collected at different time points after injury for evaluation. MicroCT and histological analyses were conducted. EdU incorporation analysis was performed to assay cell proliferation. Gli1-Cre
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8267 Corn oil, delivery vehicle for fat-soluble compounds