EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
ALT reagent with thionicotinamide adenine dinucleotide.
PMID 2776309
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7375
Thionicotinamide adenine dinucleotide, ≥90%
C21H27N7O13P2S