EMAIL THIS PAGE TO A FRIEND
European journal of ophthalmology
Novel variants of RPGR in X-linked retinitis pigmentosa families and genotype-phenotype correlation.
PMID 27768226
Abstract
To identify novel mutations in the retinitis pigmentosa GTPase regulator (RPGR) gene and retinitis pigmentosa 2 (RP2) gene underlying X-linked retinitis pigmentosa (XLRP) and assess genotype-phenotype correlations. The patient cohort, consisting of 13 individuals from 3 unrelated XLRP families, underwent comprehensive ophthalmologic examination. The open reading frames of RPGR and RP2 were analyzed with Sanger sequencing in each patient. The identified genetic variants were defined as mutations or polymorphisms on the basis of their pathological effect. We found 3 genetic variants: a novel mutation c.1591G>T in exon 14 and a novel polymorphism c.1105C>T in exon 10, resulting in p.Glu531* and p.Arg369Cys of RPGR gene, respectively, and one already known mutation c.413A>G in exon 2, resulting in a p.Glu138Gly of RP2 gene. Considering our XLRP probands, RPGR-related phenotypic damages were similar and less severe than those of the patient with the RP2 mutation. On the other hand, the female carriers of XLRP variants showed different RPGR-related consequences, ranging from rods hypofunctionality in c.1591G>T nonsense heterozygosity to no retinal changes in c.1105C>T polymorphic heterozygosity. These findings broaden the spectrum of RPGR mutations and phenotypic variability of the disease, which will be useful for genetic consultation and diagnosis in the future.