EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Combined Local Pulmonary and Systemic Delivery of AT2R Gene by Modified TAT Peptide Nanoparticles Attenuates Both Murine and Human Lung Carcinoma Xenografts in Mice.
PMID 27769520
Abstract
To evaluate the potential of cell-penetrating peptide-based delivery of apoptosis-inducer gene in cancer therapy, a modified HIV-1 TAT peptide (dimerized TAT peptide, dTAT) was studied. The dTAT and plasmid DNA (pDNA) complexes (dTAT-pDNA) were condensed using calcium chloride (dTAT-pDNA-Ca(2+)). This simple nonviral formulation approach showed high levels of gene expression inxa0vitro without any cytotoxicity. In mouse studies, a single intratracheal (IT) aerosol spray or 2 intravenous (IV) injections of the dTAT, apoptosis-inducer gene, angiotensin II type 2 receptor (AT2R), and Ca(2+) complexes (dTAT-pAT2R-Ca(2+)) significantly attenuated the acutely growing mouse Lewis lung carcinoma allografts in mouse lungs. Furthermore, single IT (pxa0= 0.054) and the combination of IT and IV (p < 0.05) administrations of dTAT-pAT2R-Ca(2+) markedly attenuated slowly growing and relatively large-sized H358 human bronchioloalveolar carcinoma xenografts in mouse lungs. These results indicate that the dTAT-pDNA-Ca(2+) effectively delivered the gene to cancer cells by either IT or IV administration although the local pulmonary delivery of the dTAT-pAT2R-Ca(2+) showed more effective growth inhibition of orthotopic lung cancer grafts. Thus, the present study offers preclinical proof of concept that a dTAT-based nonviral gene delivery method via IT administration may be an effective lung cancer gene therapy.