Clinical pharmacology and therapeutics
Circulating acetaminophen metabolites are toxicokinetic biomarkers of acute liver injury.
PMID 27770431
Acetaminophen (paracetamol-APAP) is the most common cause of drug-induced liver injury in the Western world. Reactive metabolite production by cytochrome P450 enzymes (CYP-metabolites) causes hepatotoxicity. We explored the toxicokinetics of human circulating APAP metabolites following overdose. Plasma from patients treated with acetylcysteine (NAC) for a single APAP overdose was analyzed from discovery (n = 116) and validation (n = 150) patient cohorts. In the discovery cohort, patients who developed acute liver injury (ALI) had higher CYP-metabolites than those without ALI. Receiver operator curve (ROC) analysis demonstrated that at hospital presentation CYP-metabolites were more sensitive/specific for ALI than alanine aminotransferase (ALT) activity and APAP concentration (optimal CYP-metabolite receiver operating characteristic area under the curve (ROC-AUC): 0.91 (95% confidence interval (CI) 0.83-0.98); ALT ROC-AUC: 0.67 (0.50-0.84); APAP ROC-AUC: 0.50 (0.33-0.67)). This enhanced sensitivity/specificity was replicated in the validation cohort. Circulating CYP-metabolites stratify patients by risk of liver injury prior to starting NAC. With development, APAP metabolites have potential utility in stratified trials and for refinement of clinical decision-making.