EMAIL THIS PAGE TO A FRIEND
The Journal of pathology
Fgf10 deficiency is causative for lethality in a mouse model of bronchopulmonary dysplasia.
PMID 27770432
Abstract
Inflammation-induced FGF10 protein deficiency is associated with bronchopulmonary dysplasia (BPD), a chronic lung disease of prematurely born infants characterized by arrested alveolar development. So far, experimental evidence for a direct role of FGF10 in lung disease is lacking. Using the hyperoxia-induced neonatal lung injury as a mouse model of BPD, the impact of Fgf10 deficiency in Fgf10
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0463
BPD, ≥98% (HPLC)
C17H10O5