EMAIL THIS PAGE TO A FRIEND
Immunobiology
Down-regulation of Na
PMID 27771174
Abstract
The role of Na
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0605
Z-Asp(O-Me)-Glu(O-Me)-Val-Asp(O-Me) fluoromethyl ketone, ≥90% (HPLC)
C30H41N4O12F