EMAIL THIS PAGE TO A FRIEND
Epidemiology (Cambridge, Mass.)
The Trend-in-trend Research Design for Causal Inference.
PMID 27775954
Abstract
Cohort studies can be biased by unmeasured confounding. We propose a hybrid ecologic-epidemiologic design called the trend-in-trend design, which requires a strong time trend in exposure, but is unbiased unless there are unmeasured factors affecting outcome for which there are time trends in prevalence that are correlated with time trends in exposure across strata with different exposure trends. Thus, the conditions under which the trend-in-trend study is biased are a subset of those under which a cohort study is biased. The trend-in-trend design first divides the study population into strata based on the cumulative probability of exposure given covariates, which effectively stratifies on time trend in exposure, provided there is a trend. Next, a covariates-free maximum likelihood model estimates the odds ratio (OR) using data on exposure prevalence and outcome frequency within cumulative probability of exposure strata, across multiple periods. In simulations, the trend-in-trend design produced ORs with negligible bias in the presence of unmeasured confounding. In empiric applications, trend-in-trend reproduced the known positive association between rofecoxib and myocardial infarction (observed OR: 1.2, 95% confidence interval: 1.1, 1.4), and known null associations between rofecoxib and severe hypoglycemia (OR = 1.1 [0.92, 1.3]) and nonvertebral fracture (OR = 0.84 [0.64, 1.1]). The trend-in-trend method may be useful in settings where there is a strong time trend in exposure, such as a newly approved drug or other medical intervention. See video abstract at, http://links.lww.com/EDE/B178.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0613 Rofecoxib, ≥98% (HPLC)
C17H14O4S