EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Amphetamine and derivatives in natural weight loss pills and dietary supplements by capillary electrophoresis-tandem mass spectrometry.
PMID 27776329
Abstract
A capillary electrophoresis-tandem mass spectrometry (CE-MS/MS) method for amphetamine (AM), phentermine (PTM), methamphetamine (MAM), methylenedioxyamphetamine (MDA), methylenedioxymethamphetamine (MDMA), and methylenedioxyethylamphetamine (MDEA) in commercial samples of homeopathic and phytotherapic medicines and dietary supplements is presented. The samples were submitted to a modified QuEChERS extraction procedure (at apparent pH 13) followed by electrophoretic separation in 0.1molL