EMAIL THIS PAGE TO A FRIEND
Free radical research
Plant sterol-enriched soy milk consumption modulates 5-lipoxygenase, 12-lipoxygenase, and myeloperoxidase activities in healthy adults - a randomized-controlled trial.
PMID 27776459
Abstract
A randomized, double blind, placebo-controlled and crossover study was conducted to simultaneously measure the effects, after 3-h and 4-week daily exposure to plant sterols-enriched food product, on inflammation, oxidative status, 5-lipoxygenase, 12-lipoxygenase, and myeloperoxidase activities in healthy adults. Eighteen healthy participants (67% female, 35.3 (mean) ± 9.5 (SD) years, mean body mass index 22.8 kg m-2) received two soy milk (20g) treatments daily: placebo and one containing 2.0 g free plant sterols equivalent of their palmitates (β-sitosterol, 55%; campesterol, 29%; stigmasterol, 23%). F2-isoprostanes, leukotriene B4, sitosterol, campesterol, and stigmasterol concentrations were measured in the blood plasma and urine, using stable isotope-labeled gas chromatography-mass spectrometry. High-sensitivity c-reactive protein, tumor necrosis factor-α, and lipoxin A4 concentrations in blood serum were measured using commercially available enzyme immunoassays. Myeloperoxidase activity, serum lipid hydroperoxides, plasma and urinary F2-isoprostanes, plasma and urinary leukotriene B4, and plasma high-sensitivity c-reactive protein concentrations were significantly reduced, while circulating lipoxin A4 concentrations were significantly elevated after 4-week plant sterols treatment. Plant sterols treatment decreased plasma leukotriene B4 and increased plasma lipoxin A4 concentrations acutely. Total plant sterols, β-sitosterol, campesterol, and stigmasterol concentrations were significantly elevated after 4-week treatments compared with the pre-treatment concentrations. Our results suggest that dietary plant sterols, in the combination used, can alleviate lipid peroxidation and inflammatory events in vivo. These effects are possibly exerted via the modulation of myeloperoxidase, 5-lipoxygenase, and 12-lipoxygenase activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5157
Campesterol, ~65%
C28H48O
L0517
Leukotriene B4, ~100 μg/mL in ethanol, ≥97%
C20H32O4