EMAIL THIS PAGE TO A FRIEND
Mutation research
Lanthanum nitrate genotoxicity evaluation: Ames test, mouse micronucleus assay, and chromosome aberration test.
PMID 27776686
Abstract
The increasing use of rare-earth elements (REE) and their compounds has led to their accumulation in the environment and has raised concern about their safety. The toxic effects of REE such as lanthanum are largely unknown; genotoxicity studies have been limited and results are controversial. We evaluated the genotoxicity of lanthanum nitrate (La(NO