EMAIL THIS PAGE TO A FRIEND
Pacing and clinical electrophysiology : PACE
Long-Term Stability and Clinical Utility of Amplified Atrial Electrograms in a Single-Lead ICD System with Floating Atrial Electrodes.
PMID 27778352
Abstract
Available atrial electrograms in implantable cardioverter defibrillators (ICDs) improve arrhythmia diagnosis, allow monitoring for atrial fibrillation, and may reduce the risk of inappropriate therapies. A recently introduced ICD system using a single-lead with floating atrial electrodes provides diagnostic capability of a dual-chamber system without placing an additional lead. Data on long-term clinical performance of this system are limited. We retrospectively analyzed data from 35 consecutive patients implanted with Biotronik VR-T DX devices and Linox The single-lead ICD system using a floating atrial dipole provides reliable recording of atrial signals during sinus rhythm and arrhythmias. Our data suggest that the system may offer diagnostic advantages of a dual-chamber device without potential risks of an additional atrial lead.