EMAIL THIS PAGE TO A FRIEND
Cancer research
Mechanisms through Which Hypoxia-Induced Caveolin-1 Drives Tumorigenesis and Metastasis in Hepatocellular Carcinoma.
PMID 27784747
Abstract
In solid tumors, hypoxia triggers an aberrant vasculogenesis, enhances malignant character, and elevates metastatic risk. The plasma membrane organizing protein caveolin-1 (Cav1) is increased in a variety of cancers, including hepatocellular carcinoma (HCC), where it contributes to metastatic capability. However, the reason for elevation of Cav1 in tumor cells and the mechanistic basis for its contributions to metastatic risk are not fully understood. Here, we show that in HCC cells, hypoxia elevates expression of Cav1, which then acts through the calcium-binding protein S100P to promote metastasis. Hypoxic regions of HCC xenografts displayed elevated expression of Cav1. Hypoxia promoted HCC cell migration and invasion and distant pulmonary metastases, whereas Cav1 silencing abolished these effects. Gene expression profiling revealed that hypoxia-induced Cav1 functioned as a positive regulator of S100P via activation of the NF-κB pathway. S100P elevation under hypoxic conditions was abrogated by silencing of Cav1 or NF-κB function. Conversely, restoring S100P in Cav1-silenced cells rescued the migratory potential of HCC cells along with tumor formation and lung metastasis. In clinical specimens of HCC, we observed S100P overexpression to correlate with venous invasion, microsatellites, direct liver invasion, and absence of tumor encapsulation. Collectively, our findings demonstrated how hypoxia-induced expression of Cav1 in HCC cells enhances their invasive and metastatic potential. Cancer Res; 76(24); 7242-53. ©2016 AACR.