EMAIL THIS PAGE TO A FRIEND
Lab on a chip
A benchtop capillary flow layer-by-layer (CF-LbL) platform for rapid assembly and screening of biodegradable nanolayered films.
PMID 27785506
Abstract
Capillary flow layer-by-layer (CF-LbL) is a microfluidic platform for high throughput preparation and screening of nanolayered polymer films. Using a simple benchtop version of CF-LbL, we systematically studied the effects of various flow conditions and channel geometries on the thickness and surface roughness of the resulting films. We also investigated the biocompatibility and degradation behaviors of a series of enzymatically-degradable films made from naturally derived polymers, i.e. either alginate or hyaluronic acid as the anionic species and poly-l-arginine as the positive species. Furthermore, using one optimized film formulation for coating on the inside walls of a microfluidic chip, we successfully demonstrated the ability of this film to capture and rapidly release cancer cells from whole blood. This simple platform is expected to be a powerful tool to increase the accessibility of the LbL film assembly to a broader scientific community.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56485
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)
C4H4NNaO6S