Journal of the science of food and agriculture
Effect of probiotics on patulin removal from synbiotic apple juice.
PMID 27785791
Studies have reported the occurrence of the mycotoxin patulin in apple products. The aim of this study was to produce synbiotic apple juice and investigate the detoxification of patulin by Lactobacillus acidophilus and Lactobacillus plantarum as probiotic strains. The impact of seven process variables on efficiency of toxin removal was investigated using Plackett-Burman design and presence of the surface-layer proteins as binding site of probiotics to patulin was confirmed during 6 weeks of cold storage. Results showed that the removal of patulin by probiotic bacteria from apple juice depends significantly (P < 0.05) on the fructooligosaccharide content (as a prebiotic), concentration of patulin and the addition of ascorbic acid. Sodium dodecyl sulfate polyacrylamide gel electrophoresis of cell surface proteins of probiotic strains revealed that surface layer proteins have an important role in patulin removal from apple juice. In the best conditions, 91.23% of initial patulin concentration was removed from juice during 6 weeks refrigerated storage. No significant difference was observed in organoleptic properties of the synbiotic apple juice and raw sample. In the best condition reported in this study, contaminated synbiotic apple juice by patulin will be safe for consumers after the first day of probiotic inoculation. © 2016 Society of Chemical Industry.