EMAIL THIS PAGE TO A FRIEND
Thrombosis and haemostasis
Coagulation factor XII induces pro-inflammatory cytokine responses in macrophages and promotes atherosclerosis in mice.
PMID 27786338
Abstract
Atherosclerosis is considered a chronic inflammatory disease of the vessel wall. Coagulation pathways and immune responses contribute to disease development. The role of coagulation factor XII (FXII) in vascular inflammation, however, remains controversial. We here investigated the function of FXII in atherosclerosis using apolipoprotein E and FXII-deficient (F12
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56924
Tricosanoic acid tryptamide, analytical standard
C33H56N2O