EMAIL THIS PAGE TO A FRIEND
Journal of nuclear medicine : official publication, Society of Nuclear Medicine
Preclinical Evaluation and Quantification of
PMID 27789720
Abstract
The cerebral adenosine A
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
317551
1,3-Propanediol di-p-tosylate, 98%
C17H20O6S2