EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Reactions of Trimethylaluminium: Modelling the Chemical Degradation of Synthetic Lubricants.
PMID 27792841
Abstract
In investigating and seeking to mimic the reactivity of trimethylaluminium (TMA) with synthetic, ester-based lubricating oils, the reaction of methyl propionate 1 was explored with 1, 2 and 3 equivalents of the organoaluminium reagent. Spectroscopic analysis points to the formation of the adduct 1(TMA) accompanied only by the low level 1:1 production of Me
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
108057
Methyl phenylacetate, ReagentPlus®, ≥99%
C9H10O2
109258
Methyl propionate, 99%
C4H8O2