EMAIL THIS PAGE TO A FRIEND
British journal of pharmacology
Hydrogen peroxide preferentially activates capsaicin-sensitive high threshold afferents via TRPA1 channels in the guinea pig bladder.
PMID 27792844
Abstract
There is increasing evidence suggesting that ROS play a major pathological role in bladder dysfunction induced by bladder inflammation and/or obstruction. The aim of this study was to determine the effect of H 'Close-to-target' single unit extracellular recordings were made from fine branches of pelvic nerves entering the guinea pig bladder, in flat sheet preparations, in vitro. H The findings show that H
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS002866
1-Piperazinecarboxamide, AldrichCPR
C5H11N3O
C191
Capsazepine, ≥98% (HPLC), solid
C19H21ClN2O2S