EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Antibacterial profiling of abietane-type diterpenoids.
PMID 27793449
Abstract
Abietic and dehydroabietic acid are interesting diterpenes with a highly diverse repertoire of associated bioactivities. They have, among others, shown antibacterial and antifungal activity, potentially valuable in the struggle against the increasing antimicrobial resistance and imminent antibiotic shortage. In this paper, we describe the synthesis of a set of 9 abietic and dehydroabietic acid derivatives containing amino acid side chains and their in vitro antimicrobial profiling against a panel of human pathogenic microbial strains. Furthermore, their in vitro cytotoxicity against mammalian cells was evaluated. The experimental results showed that the most promising compound was 10 [methyl N-(abiet-8,11,13-trien-18-yl)-d-serinate], with an MIC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00089
Dehydroabietic acid, ≥95% (LC/MS-ELSD)
C20H28O2