EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Semi-preparative HPLC separation followed by HPLC/UV and tandem mass spectrometric analysis of phorbol esters in Jatropha seed.
PMID 27794223
Abstract
Phorbol esters (PEs) are well known as the main toxic compounds in Jatropha curcas Linnaeus (JCL), the seed oil of which has been considered as a major feedstock for the production of biodiesel. In the present study, we investigated a series of PEs extracted from JCL seed kernels with methanol (MeOH), and identified more than seven components contained in the PEs. The isolation of main five components of a series of PEs was revised using a semi-preparative reversed phase HPLC analysis of ODS-3 column. The five peaks of components were successfully isolated, and peaks of J2, J3, J5, and J7 were assigned to be Jatropha factors C
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
723134
Tris(2-pyridylmethyl)amine, 98%
C18H18N4