EMAIL THIS PAGE TO A FRIEND
Experimental neurology
Neuroprotective effect of lercanidipine in middle cerebral artery occlusion model of stroke in rats.
PMID 27794423
Abstract
Oxidative stress, inflammation and apoptotic neuronal cell death are cardinal mechanisms involved in the cascade of acute ischemic stroke. Lercanidipine apart from calcium channel blocking activity possesses anti-oxidant, anti-inflammatory and anti-apoptotic properties. In the present study, we investigated neuroprotective efficacy and therapeutic time window of lercanidipine in a 2h middle cerebral artery occlusion (MCAo) model in male Wistar rats. The study design included: acute (pre-treatment and post-treatment) and sub-acute studies. In acute studies (pre-treatment) lercanidipine (0.25, 0.5 and 1mg/kg, i.p.) was administered 60min prior MCAo. The rats were assessed 24h post-MCAo for neurological deficit score (NDS), motor deficit paradigms (grip test and rota rod) and cerebral infarction via 2,3,5-triphenyltetrazolium chloride (TTC) staining. The most effective dose was found to be at 0.5mg/kg, i.p., which was considered for further studies. Regional cerebral blood flow (rCBF) was monitored till 120min post-reperfusion to assess vasodilatory property of lercanidipine (0.5mg/kg, i.p.) administered at two different time points: 60min post-MCAo and 15min post-reperfusion. In acute studies (post-treatment) lercanidipine (0.5mg/kg, i.p.) was administered 15min, 120min and 240min post-reperfusion. Based on NDS and cerebral infarction via TTC staining assessed 24h post-MCAo, effectiveness was evident upto 120min. For sub-acute studies same dose/vehicle was repeated for next 3days and magnetic resonance imaging (MRI) was performed 96h after the last dose. Biochemical markers estimated in rat brain cortex 24h post-MCAo were oxidative stress (malondialdehyde, reduced glutathione, nitric oxide, superoxide dismutase), blood brain barrier damage (matrix metalloproteinases-2 and -9) and apoptotic (caspase-3 and -9). Lercanidipine significantly reduced NDS, motor deficits and cerebral infarction volume as compared to the control group. Lercanidipine (60min post-MCAo) significantly increased rCBF (86%) as compared to vehicle treated MCAo group (64%) 120min post-reperfusion, but failed to show vasodilatation with 15min post-reperfusion group. Lercanidipine (13.78±2.78%) significantly attenuated percentage infarct volume as evident from diffusion-weighted (DWI) and T2-weighted images as compared to vehicle treated MCAo group (25.90±2.44%) investigated 96h post-MCAo. The apparent diffusion coefficient (ADC) was also significantly improved in lercanidipine group as compared to control group. Biochemical alterations were significantly ameliorated by lercanidipine till 120min post-reperfusion group and MMP-9 inhibition observed even with 240min group. Thus, lercanidipine revealed significant neuroprotective effect mediated through attenuation of oxidative stress, inflammation and apoptosis.