EMAIL THIS PAGE TO A FRIEND
Glia
Endogenous GABA controls oligodendrocyte lineage cell number, myelination, and CNS internode length.
PMID 27796063
Abstract
Adjusting the thickness and internodal length of the myelin sheath is a mechanism for tuning the conduction velocity of axons to match computational needs. Interactions between oligodendrocyte precursor cells (OPCs) and developing axons regulate the formation of myelin around axons. We now show, using organotypic cerebral cortex slices from mice expressing eGFP in Sox10-positive oligodendrocytes, that endogenously released GABA, acting on GABA