EMAIL THIS PAGE TO A FRIEND
Diabetologia
Insulitis in human diabetes: a histological evaluation of donor pancreases.
PMID 27796420
Abstract
According to the consensus criteria developed for type 1 diabetes, an individual can be diagnosed with insulitis when ≥ 15 CD45 Insulitis was determined by examining CD45 Overall, 28% of the type 2 diabetic donors fulfilled the consensus criteria for insulitis developed for type 1 diabetes. Of the type 1 diabetic donors, 31% fulfilled the criteria. None of the non-diabetic donors met the criteria. Only type 1 diabetic donors had ≥ 15 CD3 The current definition of insulitis cannot be used to distinguish pancreases retrieved from individuals with type 1 diabetes from those with type 2 diabetes. On the basis of our findings, we propose a revised definition of insulitis, with a positive diagnosis when ≥ 15 CD3