EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Growth of four tropical tree species in petroleum-contaminated soil and effects of crude oil contamination.
PMID 27796985
Abstract
Under greenhouse conditions, we evaluated establishment of four tree species and their capacity to degrade crude oil recently incorporated into the soil; the species were as follows: Cedrela odorata (tropical cedar), Haematoxylum campechianum (tinto bush), Swietenia macrophylla (mahogany), and Tabebuia rosea (macuilis). Three-month-old plants were planted in soil with three treatments of heavy petroleum and a control (C0 0 mg kg