EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
The A-type potassium current: methoxybenzenes increase the rate of inactivation in snail neurons.
PMID 2779844
Abstract
The effects of anisole (methoxybenzene) and veratrole (1,2-dimethoxybenzene) on the resting membrane potential (RMP), action potential (AP) and membrane potassium currents were investigated in snail neurons under current- and voltage-clamp conditions. Anisole (0.9-3 mM) scarcely influenced the AP, but it moderately depolarized the membrane and increased the neuronal excitability. Anisole accelerated the time-dependent inactivation of the A-current and decreased the time constant of the decay in a dose-dependent way (K1/2 = 1.2 mM). The potential-dependence of the steady-state activation or inactivation of the A-current was not influenced by anisole. An analysis of the Hill plot of anisole action on the time constant of decay of the A-current showed a one-to-two binding stoichiometry (nH = 2.1). Veratrole (2-8 mM) was effective similarly to anisole on the A-current, but proved less potent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
140155
1,2-Dimethoxybenzene, ReagentPlus®, 99%
C8H10O2
W379905
1,2-Dimethoxybenzene, ≥99%, FG
C8H10O2