EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Residues of organochlorine pesticides in surface water of a megacity in central China: seasonal-spatial distribution and fate in Wuhan.
PMID 27798806
Abstract
Surface water quality closely correlating with human health suffered increasing organochlorine pesticide (OCP) pollution due to the intensive anthropogenic activities in megacities. In the present study, 112 water samples collected from 14 lakes and 11 drinking water source sites in Wuhan were detected for the residues of OCPs in November 2013 and July 2014, respectively. The ΣOCPs ranged from 5.61 to 13.62 ng L
Related Materials