EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
Msi RNA-binding proteins control reserve intestinal stem cell quiescence.
PMID 27799368
Abstract
Regeneration of the intestinal epithelium is driven by multiple intestinal stem cell (ISC) types, including an active, radiosensitive Wnt
Related Materials