Diagnostic and interventional radiology (Ankara, Turkey)
Endovascular treatment of iliofemoral deep vein thrombosis in pregnancy using US-guided percutaneous aspiration thrombectomy.
PMID 27801353
We aimed to describe ultrasonography (US)-guided percutaneous aspiration thrombectomy in pregnant women with iliofemoral deep vein thrombosis. This study included nine pregnant women with acute and subacute iliofemoral deep vein thrombosis, who were severe symptomatic cases with massive swelling and pain of the leg. Patients were excluded from the study if they had only femoropopliteal deep vein thrombosis or mild symptoms of deep vein thrombosis. US-guided percutaneous aspiration thrombectomy was applied to achieve thrombus removal and uninterrupted venous flow. The treatment was considered successful if there was adequate venous patency and symptomatic relief. Complete or significant thrombus removal and uninterrupted venous flow from the puncture site up to the iliac veins were achieved in all patients at first intervention. Complete relief of leg pain was achieved immediately in seven patients (77.8%). Two patients (22.2%) had a recurrence of thrombosis in the first week postintervention. One of them underwent a second intervention, where percutaneous aspiration thrombectomy was performed again with successful removal of thrombus and establishment of in line flow. Two patients were lost to follow-up after birth. None of the remaining seven patients had rethrombosis throughout the postpartum period. Symptomatic relief was detected clinically in these patients. Endovascular treatment with US-guided percutaneous aspiration thrombectomy can be considered as a safe and effective way to remove thrombus from the deep veins in pregnant women with acute and subacute iliofemoral deep vein thrombosis.