EMAIL THIS PAGE TO A FRIEND
Journal of nuclear cardiology : official publication of the American Society of Nuclear Cardiology
The impact of combination of regadenoson and isometric exercise on image quality of myocardial perfusion scintigraphy.
PMID 27804069
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
716707
Cadmium zinc telluride, 99.9999% trace metals basis
Cd0.9Zn0.1Te