EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
Weakening Circular Dichroism of Plasmonic Nanospirals Induced by Surface Grafting with Alkyl Ligands.
PMID 27805771
Abstract
Silver nanospirals with strong chiroptical activity (characterized by circular dichroism) are grafted with achiral alkyl ligands, resulting in a weakening of the chiroptical activity. The chiroptical weakening is exacerbated with increasing bond energy of the Ag-ligand contacts, which is ascribed to the effective medium screening effect and electron withdrawal toward the alkyl ligands.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
471836
1-Octanethiol, ≥98.5%
C8H18S
N5502
Nonanoic acid, ≥97%
C9H18O2
O5802
Octylamine, 99%
C8H19N