EMAIL THIS PAGE TO A FRIEND
Planta medica
Chlorogenic Compounds from Coffee Beans Exert Activity against Respiratory Viruses.
PMID 27806409
Abstract
Chlorogenic acids are secondary metabolites in diverse plants. Some chlorogenic acids extracted from traditional medicinal plants are known for their healing properties, e.g., against viral infections. Also, green coffee beans are a rich source of chlorogenic acids, with 5-