EMAIL THIS PAGE TO A FRIEND
Progress in clinical and biological research
Influence of lidocaine on plasma protein binding and pharmacokinetics of verapamil in dogs.
PMID 2780641
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
130591
Tris(2-butoxyethyl) phosphate, 94%
C18H39O7P