EMAIL THIS PAGE TO A FRIEND
The Journal of experimental medicine
Mafb lineage tracing to distinguish macrophages from other immune lineages reveals dual identity of Langerhans cells.
PMID 27810926
Abstract
Current systems for conditional gene deletion within mouse macrophage lineages are limited by ectopic activity or low efficiency. In this study, we generated a Mafb-driven Cre strain to determine whether any dendritic cells (DCs) identified by Zbtb46-GFP expression originate from a Mafb-expressing population. Lineage tracing distinguished macrophages from classical DCs, neutrophils, and B cells in all organs examined. At steady state, Langerhans cells (LCs) were lineage traced but also expressed Zbtb46-GFP, a phenotype not observed in any other population. After exposure to house dust mite antigen, Zbtb46-negative CD64
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4937 Percoll®, pH 8.5-9.5 (25 °C), cell culture tested