EMAIL THIS PAGE TO A FRIEND
Journal of photochemistry and photobiology. B, Biology
Synthesis, characterization and anticancer effect of the ruthenium (II) polypyridyl complexes on HepG2 cells.
PMID 27816647
Abstract
As one of the major cell regulated center, mitochondria are closely associated with cell proliferation, apoptosis of tumor cell. In this work, four new ruthenium (II) polypyridyl complexes [Ru(bpy)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295272
1,1-Difluoroethylene, ≥99%
C2H2F2