EMAIL THIS PAGE TO A FRIEND
Medical engineering & physics
An active simulator for neonatal intubation: Design, development and assessment.
PMID 27818075
Abstract
This study describes the technical realization and the pre-clinical validation of a instrumented neonatal intubation skill trainer able to provide objective feedback for the improvement of clinical competences required for such a delicate procedure. The Laerdal