EMAIL THIS PAGE TO A FRIEND
Platelets
The effects of hydrogen sulfide on platelet-leukocyte aggregation and microvascular thrombolysis.
PMID 27819526
Abstract
The volatile transmitter hydrogen sulfide (H