EMAIL THIS PAGE TO A FRIEND
Clinical nuclear medicine
Investigating the Chemokine Receptor 4 as Potential Theranostic Target in Adrenocortical Cancer Patients.
PMID 27819856
Abstract
Adrenocortical carcinoma (ACC) is a rare but aggressive endocrine tumor with limited treatment options. Preclinical studies confirmed overexpression of the chemokine receptor 4 (CXCR4) in this cancer type. This study aimed to analyze the role of CXCR4 imaging using Ga-pentixafor for ACC staging and selection of patients for CXCR4-directed endoradiotherapy. Thirty patients with histologically proven advanced, metastasized ACC underwent F-FDG PET/CT and Ga-pentixafor PET/CT within a time interval of 3 ± 4 days to evaluate suitability for CXCR4-directed endoradiotherapy. Scans were analyzed retrospectively for visual extent of ACC and SUVmax/mean of the tumor lesions. Ga-pentixafor PET was compared with F-FDG PET, the reference imaging standard. All patients were rated for suitability of CXCR4-directed endoradiotherapy considering patient's history, previous treatment, and CXCR4 expression of FDG-positive lesions compared with background activity within the same organ. All patients had lesions that were positive for both F-FDG and Ga-pentixafor PET and were rated as positive for disease. In 2 patients (7%), Ga-pentixafor PET identified more lesions compared with F-FDG PET. In 5 patients (17%) and 10 patients (33%), complementary and comparable information, respectively, was provided by dual-tracer imaging. In 13 patients (43%), more tumor lesions were identified by F-FDG PET compared with Ga-pentixafor PET. The F-FDG uptake of the malignant lesions was significantly higher (P < 0.01) than the SUVmax/mean for Ga-pentixafor. Overall, 70% of the patients were rated as suitable or potentially suitable for CXCR4-directed treatment. Ga-pentixafor allows in vivo imaging of CXCR4 expression in patients with advanced ACC and may serve as companion diagnostic tool in selecting patients for potential CXCR4-directed endoradiotherapy. Seventy percent of the patients with advanced, metastasized ACC may be suitable for a CXCR4-directed treatment after failure of standard treatment options.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G8022
Gastrin Releasing Peptide human, ≥97% (HPLC)
C130H204N38O31S2