EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Fate of the
PMID 27820683
Abstract
The fate of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31141
Prosulfocarb, PESTANAL®, analytical standard
C14H21NOS