EMAIL THIS PAGE TO A FRIEND
Supportive care in cancer : official journal of the Multinational Association of Supportive Care in Cancer
Psychosocial concerns and needs of cancer survivors treated at a comprehensive cancer center and a community safety net hospital.
PMID 27822710
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
248401
Magnesium methyl carbonate solution, 2.0 M in DMF
C3H6MgO4