EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Bis(phenylsulfone) as a strong electron acceptor of thermally activated delayed fluorescent emitters.
PMID 27824181
Abstract
Bis(phenylsulfone) was developed as a strong electron acceptor of thermally activated delayed fluorescent emitters. The connection of two electron withdrawing phenylsulfone moieties through meta-position of phenyl produced the bis(phenylsulfone) acceptor and the strong electron acceptor strength of bis(phenylsulfone) enabled preparation of sky-blue and green thermally activated delayed fluorescent emitters in combination with weak carbazole donors. The bis(phenylsulfone) acceptor and carbazole donor combined organic materials performed as thermally activated delayed fluorescent emitters with a high quantum efficiency of 18.3%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
757322
3,6-Di-tert-butylcarbazole, 97%
C20H25N