EMAIL THIS PAGE TO A FRIEND
Biosensors & bioelectronics
Paper-based microfluidic biofuel cell operating under glucose concentrations within physiological range.
PMID 27825524
Abstract
This work addresses the development of a compact paper-based enzymatic microfluidic glucose/O
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS006368
diglycidyl ether, AldrichCPR
C6H10O3