EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Quantification of suvorexant in urine using gas chromatography/mass spectrometry.
PMID 27825624
Abstract
Suvorexant (Belsomra
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
287636
(1S,2S)-(+)-Pseudoephedrine, 98%
C10H15NO
C8912
Cocaine free base
C17H21NO4
Z103
Zolpidem, ≥98% (HPLC), solid
C19H21N3O