EMAIL THIS PAGE TO A FRIEND
Behavioural brain research
Activation of mu-opioid receptors in the ventral pallidum decreases the negative hedonic evaluation of a conditioned aversive taste in rats.
PMID 27825896
Abstract
Conditioned taste aversion (CTA) causes a shift in the hedonic evaluation of a conditioned stimulus (CS) from positive to negative, and reduces the CS intake. Mu-opioid receptors (MORs) in the ventral pallidum (VP) are known to be involved in the hedonic evaluation of positive rewarding stimuli; however, their involvement in evaluation of a negative aversive stimulus is still unclear. To explore the neural mechanisms of the negative hedonic evaluation of the CS in CTA, we examined the effects of the activation of VP MORs on the behavioral responses of rats to a CS. Rats implanted with guide cannulae into the bilateral VP received a pairing of 5mM saccharin solution as a CS with an intraperitoneal injection of 0.15M lithium chloride as an unconditioned stimulus. On the test day, after microinjections of MOR agonist [D-Ala