EMAIL THIS PAGE TO A FRIEND
Diabetologia
A Tbc1d1
PMID 27826658
Abstract
TBC1D1 (tre-2/USP6, BUB2, cdc16 domain family member 1) is a Rab GTPase-activating protein (RabGAP) that has been implicated in regulating GLUT4 trafficking. TBC1D1 can be phosphorylated by the AMP-activated protein kinase (AMPK) on Ser Whole-body glucose homeostasis and muscle glucose uptake were assayed in mice bearing a Tbc1d1 Skeletal muscle-specific deletion of Ampkα1/a2 in mice prevented AICAR-induced hypoglycaemia and muscle glucose uptake. The Tbc1d1 TBC1D1-Ser
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9978
AICAR, ≥98% (HPLC), powder
C9H14N4O5