Applied biochemistry and biotechnology
Use of Bacillus amyloliquefaciens HZ-12 for High-Level Production of the Blood Glucose Lowering Compound, 1-Deoxynojirimycin (DNJ), and Nutraceutical Enriched Soybeans via Fermentation.
PMID 27826807
1-Deoxynojirimycin (DNJ) is an efficient α-glucosidase inhibitor (α-GI) with potential applications in the prevention and treatment of diabetes. In this study, 16 Bacillus strains were screened for α-GI rate, and the strain HZ-12 with the highest α-GI rate was identified as Bacillus amyloliquefaciens through the analysis of physiological biochemical characteristics and 16S rDNA sequence. By LC-MS/Q-TOF analysis, the α-GI component produced by B. amyloliquefaciens HZ-12 was identified as DNJ. Soybean was used as the substrate for the solid-state fermentation; 870xa0mg/kg DNJ was produced by B. amyloliquefaciens HZ-12 after optimizing the fermentation conditions and media, which was 3.83-fold higher than the initial yield. Also, evaluations of nutraceutical enrichment in the form of anticoagulant activity, antioxidant activity, total nitrogen (TN), and total reducing sugars (TRS) of the B. amyloliquefaciens HZ-12 fermented soybeans were substantially higher than unfermented soybeans. This study provided a promising strain for high-level production of DNJ and produced nutraceutical enriched soybeans by fermentation.