EMAIL THIS PAGE TO A FRIEND
Scientific reports
Potent Inducers of Endogenous Antimicrobial Peptides for Host Directed Therapy of Infections.
PMID 27827460
Abstract
A new concept for treatment of infections is induction of our own antimicrobial peptides and the presented novel class of inducer, aroylated phenylenediamines (APDs), gives up to 20 to 30-fold induction of the human antimicrobial peptide LL-37, in vitro. In addition, oral administration of an APD in a rabbit model of Shigellosis resulted in recovery from the infection in a few days implying that APD's are promising candidates for treatment of infections.